Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DCAF12L2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA561855
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA561855 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA561855 Supplier Invitrogen™ Supplier No. PA561855
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GLQGFEGELR GYAVQRLPEL LTERQLDLGT Highest antigen sequence identity to the following orthologs: Mouse - 73%, Rat - 73%.

This gene encodes a member of the WD repeat protein family. WD repeats are minimally conserved regions of approximately 40 amino acids typically bracketed by Gly-His and Trp-Asp (GH-WD), which may facilitate formation of heterotrimeric or multi-protein complexes. Members of this family are involved in a variety of cellular processes, including cell cycle progression, signal transduction, apoptosis, and gene regulation. This gene appears to represent an intronless retrocopy of a related multi-exon gene located on chromosome 9. However, the CDS of this intronless gene remains intact, it is conserved in other mammalian species, it is known to be transcribed, and it is therefore thought to encode a functional protein. [provided by RefSeq, May 2010].
TRUSTED_SUSTAINABILITY

Specifications

Antigen DCAF12L2
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene DCAF12L2
Gene Accession No. Q5VW00
Gene Alias DCAF12L2; DDB1 and CUL4 associated factor 12 like 2; DDB1 and CUL4 associated factor 12-like 2; DDB1- and CUL4-associated factor 12-like protein 2; WD repeat-containing protein 40C; WDR40C
Gene Symbols DCAF12L2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human DCAF12L2. Recombinant protein control fragment (Product #RP-99273).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 340578
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.