Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DCAF13 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501989038
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-9038 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-9038 Supplier Bioss Supplier No. BS9395R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Possible role in ribosomal RNA processing. May function as a substrate receptor for CUL4-DDB1 E3 ubiquitin-protein ligase complex.

Specifications

Antigen DCAF13
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene DCAF13
Gene Accession No. Q9NV06
Gene Alias DCAF13; DDB1 and CUL4 associated factor 13; DDB1- and CUL4-associated factor 13; Gm83; HSPC064; WD repeat and SOF domain-containing protein 1; WD repeats and SOF domain containing 1; WD repeats and SOF1 domain containing; Wdsof1
Gene Symbols DCAF13
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human DCAF13.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25879
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.