Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DCAF17 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA563056
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA563056 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA563056 Supplier Invitrogen™ Supplier No. PA563056
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EQETFKIVDY EDELDLLSVV AVTQIDAEGK AHLDFHCNEY GTLLKSIPLV ESWDVTYSHE VYFDRDLVLH IEQKPNRVFS CYVYQMICD Highest antigen sequence identity to the following orthologs: Mouse - 99%, Rat - 97%.

TRUSTED_SUSTAINABILITY

Specifications

Antigen DCAF17
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene DCAF17
Gene Accession No. Q5H9S7
Gene Alias 2810055O12Rik; 4833418A01Rik; A030004A10Rik; A930009G19Rik; AI448937; C2orf37; DCAF17; DDB1 and CUL4 associated factor 17; DDB1- and CUL4-associated factor 17; RGD1565551
Gene Symbols DCAF17
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human DCAF17. Recombinant protein control fragment (Product #RP-99629).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80067
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.