Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ DCAF17 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5109620
Description
Immunogen sequence: LYSFQTIAEQ FMQQKLDLGC ACRWGGTTGT VGEAPFGIPC NIKITDMPPL LFEVSSLENA FQIGGHPWHY IVTPNKKKQK GVFHICALKD NSLAKNGIQE M.
Specifications
| DCAF17 | |
| Polyclonal | |
| Unconjugated | |
| DCAF17 | |
| 2810055O12Rik; 4833418A01Rik; A030004A10Rik; A930009G19Rik; AI448937; C2orf37; DCAF17; DDB1 and CUL4 associated factor 17; DDB1- and CUL4-associated factor 17; RGD1565551 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 499807 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 1.75 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| B1H299 | |
| DCAF17 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 235-335 of human DCAF17 (NP_079276.2). | |
| 100 μL | |
| Primary | |
| Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction