Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DCAF4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5109621
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5109621 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5109621 Supplier Invitrogen™ Supplier No. PA5109621
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MNKSRWQSRR RHGRRSHQQN PWFRLRDSED RSDSRAAQPA HDSGHGDDES PSTSSGTAGT SSVPELPGFY FDPEKKRYFR LLPGHNNCNP LTKESIRQKE MESKRLRLLQ EEDRRKKIAR.

The nuclear protein p100 is a proliferation-associated protein whose expression is restricted to cell cycle phases S, G2, and M. Exclusively expressed in proliferating cells from the transition G1/S until the end of cytokinesis. During mitosis it is strictly associated with the spindle pole and with the mitotic spindle, whereas during S and G2, it is diffusely distributed throughout the nucleus. The full-length cDNA encodes a 747-amino acid protein with a putative ATP/GTP-binding site motif. RT-PCR analysis demonstrated strong expression of in lung carcinoma cell lines but not in normal lung tissues. Expression was also found in adult placenta, skeletal muscle, thymus, testis, and small intestine and in fetal brain, liver, and kidney. P100 is also correlated to cancer prognosis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DCAF4
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.94 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene DCAF4
Gene Accession No. Q8WV16
Gene Alias 1110018E21Rik; Dcaf4; DDB1 and CUL4 associated factor 4; DDB1- and CUL4-associated factor 4; DKFZp434K114; WD repeat domain 21; WD repeat domain 21A; WD repeat-containing protein 21A; WDR21; WDR21A
Gene Symbols DCAF4
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-120 of human DCAF4 (NP_056419.2).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26094
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.