Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DCAF6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA589724
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA589724 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA589724 Supplier Invitrogen™ Supplier No. PA589724
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DNNNEKLSPK PGTGEPVLSL HYSTEGTTTS TIKLNFTDEW SSIASSSRGI GSHCKSEGQE ESFVPQSSVQ PPEGDSETKA PEESSEDVTK YQEGVSAENP VENHINITQS DKFTAKPLDS NSGERNDLNL DRSCGVPEES ASSEKAKEPE TSDQTSTESA TNENNTNPEP QFQTEATGPS AHEETSTRDS A; Positive Samples: HeLa, LO2; Cellular Location: Nucleus.

NRIP is a novel 860a.a nuclear protein consisting of seven conserved WD40 domains and one NLS motif. It binds to androgen and glucocorticoid receptors and up regulates their transcriptional activity, thereby functioning as a nuclear receptor coactivator. Role of NRIP has been implicated in cell growth and also in cervical and prostrate cancer thus indicating a therapeutic intervention. Northern Blot analysis detected a high expression of NRIP in skeletal muscle and testis and low expression in heart, prostrate and adrenal gland.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DCAF6
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.36 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene DCAF6
Gene Accession No. Q58WW2
Gene Alias 1200006M05Rik; Androgen receptor complex-associated protein; ARCAP; Dcaf6; DDB1 and CUL4 associated factor 6; DDB1- and CUL4-associated factor 6; IQ motif and WD repeat-containing protein 1; IQ motif and WD repeats 1; IQWD1; MSTP055; NRIP; nuclear receptor interaction protein; PC326; RGD1561961
Gene Symbols DCAF6
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 460-650 of human IQWD1 (NP_0010179771).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55827
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.