Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DCAF6 Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA141255

Catalog No. 01-675-601


Only null left
Add to Cart

Description

Description

Suggested positive control: human skeletal muscle protein or tissue.

NRIP is a novel 860a.a nuclear protein consisting of seven conserved WD40 domains and one NLS motif. It binds to androgen and glucocorticoid receptors and up regulates their transcriptional activity, thereby functioning as a nuclear receptor coactivator. Role of NRIP has been implicated in cell growth and also in cervical and prostrate cancer thus indicating a therapeutic intervention. Northern Blot analysis detected a high expression of NRIP in skeletal muscle and testis and low expression in heart, prostrate and adrenal gland.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

DCAF6
Polyclonal
Unconjugated
DCAF6
1200006M05Rik; Androgen receptor complex-associated protein; ARCAP; Dcaf6; DDB1 and CUL4 associated factor 6; DDB1- and CUL4-associated factor 6; IQ motif and WD repeat-containing protein 1; IQ motif and WD repeats 1; IQWD1; MSTP055; NRIP; nuclear receptor interaction protein; PC326; RGD1561961
Rabbit
Antigen affinity chromatography
RUO
289181, 480085, 55827, 74106
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Western Blot
0.5 mg/mL
PBS with 0.05% BSA and 0.05% sodium azide
Q58WW2, Q9DC22
DCAF6
Synthetic peptide corresponding to residues800-850 of humanNRIP.
100 μg
Primary
Human, Mouse, Rat, Canine
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.