Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DCAF6 Polyclonal Antibody

DCAF6 Polyclonal Antibody

Supplier:  Thermo Scientific PA528335

Catalog No. PIPA528335


Only null left
Add to Cart

Description

Description

DCAF6 Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY
Specifications

Specifications

DCAF6
Polyclonal
Unconjugated
DCAF6
1200006M05Rik; Androgen receptor complex-associated protein; ARCAP; Dcaf6; DDB1 and CUL4 associated factor 6; DDB1- and CUL4-associated factor 6; IQ motif and WD repeat-containing protein 1; IQ motif and WD repeats 1; IQWD1; MSTP055; NRIP; nuclear receptor interaction protein; PC326; RGD1561961
Rabbit
Antigen Affinity Chromatography
RUO
55827
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Western Blot
1 mg/mL
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Q58WW2
DCAF6
Recombinant protein fragment corresponding to a region within amino acids 378 and 667 of Human NRIP.
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

WARNING: Cancer - www.P65Warnings.ca.gov
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.