Learn More
Invitrogen™ DCAF7 Polyclonal Antibody

Description
This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 100%.
Specifications
Specifications
| Antigen | DCAF7 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 2% sucrose and 0.09% sodium azide |
| Gene | DCAF7 |
| Gene Accession No. | P61963 |
| Gene Alias | 1700012F10Rik; 2610037L01Rik; AN11; AN11 homolog; C86529; DCAF7; DDB1 and CUL4 associated factor 7; DDB1- and CUL4-associated factor 7; dirty south; dys; fj83g06; HAN11; han11l; human anthocyanin; RGD1305140; seven-WD-repeat protein of the AN11 family-1; SWAN-1; Unknown (protein for MGC:134591); WD repeat domain 68; WD repeat-containing protein 68; WD repeat-containing protein An11 homolog; wdr68; WD-repeat protein; wu:fj83g06; zgc:63940 |
| Gene Symbols | DCAF7 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.