Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DCAF7 Recombinant Rabbit Monoclonal Antibody (JE61-64)
GREENER_CHOICE

Catalog No. PIMA544858
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA544858 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA544858 Supplier Invitrogen™ Supplier No. MA544858
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

DCAF7 Recombinant Monoclonal Antibody for Western Blot, IHC (P)

WD-repeats are motifs that are found in a variety of proteins and are characterized by a conserved core of 40-60 amino acids that commonly form a tertiary propeller structure. While proteins that contain WD-repeats participate in a wide range of cellular functions, they are generally involved in regulatory mechanisms concerning chromatin assembly, cell cycle control, signal transduction, RNA processing, apoptosis and vesicular trafficking. WDR68 (WD repeat-containing protein 68), also known as DCAF7 (DDB1- and CUL4- associated factor 7), HAN11 or WDR68, is a 342 amino acid protein that belongs to the WD repeat DCAF7 family and contains 4 WD repeats. Localizing to the cytoplasm and nucleus, WDR68 is involved in protein modification and ubiquitination pathways, and interacts directly with Dyrk1A, Dyrk1B and Dia 1. WDR68 is involved in craniofacial development, and may be involved in skin development.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DCAF7
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Recombinant Monoclonal
Clone JE61-64
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
Gene DCAF7
Gene Accession No. P61962, P61963
Gene Alias 1700012F10Rik; 2610037L01Rik; AN11; AN11 homolog; C86529; DCAF7; DDB1 and CUL4 associated factor 7; DDB1- and CUL4-associated factor 7; dirty south; dys; fj83g06; HAN11; han11l; human anthocyanin; RGD1305140; seven-WD-repeat protein of the AN11 family-1; SWAN-1; Unknown (protein for MGC:134591); WD repeat domain 68; WD repeat-containing protein 68; WD repeat-containing protein An11 homolog; wdr68; WD-repeat protein; wu:fj83g06; zgc:63940
Gene Symbols DCAF7
Host Species Rabbit
Immunogen Synthetic peptide within human WDR68 aa 199-248/342.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10238, 303602, 71833
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.