Learn More
Invitrogen™ DCAF8 Monoclonal Antibody (4G8)

Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA524563
Description
Peptide Sequence: MSSKGSSTDG RTDLANGSLS SSPEEMSGAE EGRETSSGIE VEASDLSLSL TGDDGGPNRT STESRGTETE SSGEDKDSDS MEDTGHYSIN DENRVHDRSE EEEEEEEEEE EEQPRRRVQR KRANRDQDSS DDERALEDWV SSETSALPRP RWQALPALRE RELGSSARFV YEACGARVFV QRFRLQHGLE GHTGCVNTLH FNQRGTWLAS GSDDLKVVVW DWVRRQPVLD FESGHKSNVF QVRQGSIIAT ERIRHELEKS ELQHGLGADS ELL.
This gene encodes a WD repeat-containing protein that interacts with the Cul4-Ddb1 E3 ligase macromolecular complex. Multiple alternatively spliced transcript variants have been found for this gene.
Specifications
| DCAF8 | |
| Monoclonal | |
| 1.0 mg/mL | |
| PBS with no preservative; pH 7.4 | |
| Q5TAQ9 | |
| DCAF8 | |
| WDR42A (AAH13107, 1 a.a. ~ 273 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a κ |
| ELISA, Western Blot, Immunocytochemistry | |
| 4G8 | |
| Unconjugated | |
| DCAF8 | |
| AA408877; AA433120; AA792214; AU018973; C76055; D1Dau35e; D1Ucla4; Dcaf8; DDB1 and CUL4 associated factor 8; DDB1- and CUL4-associated factor 8; GAN2; H326; WD repeat domain 42A; WD repeat-containing protein 42A; Wdr42a | |
| Mouse | |
| Protein A | |
| RUO | |
| 50717 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.