Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DCAF8 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA555594
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA555594 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA555594 Supplier Invitrogen™ Supplier No. PA555594
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MSSKGSSTDG RTDLANGSLS SSPEEMSGAE EGRETSSGIE VEASDLSLSL TGDDGGPNRT STESRGTDTE SSG Highest antigen sequence identity to the following orthologs: Mouse - 90%, Rat - 96%.

This gene encodes a WD repeat-containing protein that interacts with the Cul4-Ddb1 E3 ligase macromolecular complex. Multiple alternatively spliced transcript variants have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DCAF8
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene DCAF8
Gene Accession No. Q5TAQ9
Gene Alias AA408877; AA433120; AA792214; AU018973; C76055; D1Dau35e; D1Ucla4; Dcaf8; DDB1 and CUL4 associated factor 8; DDB1- and CUL4-associated factor 8; GAN2; H326; WD repeat domain 42A; WD repeat-containing protein 42A; Wdr42a
Gene Symbols DCAF8
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human DCAF8. Recombinant protein control fragment (Product #RP-94197).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 50717
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.