Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DCAMKL2 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP177128
Description
DCAMKL2 Polyclonal specifically detects DCAMKL2 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
DCAMKL2 | |
Polyclonal | |
Unconjugated | |
PBS with 0.02% Sodium Azide | |
CL2EC 2.7.11.1, CLICK-II, CLIK2, DCDC3, DCDC3BCaMK-like CREB regulatory kinase 2, DCK2DCAMKL2, DKFZp761I032, doublecortin and CaM kinase-like 2, Doublecortin domain-containing protein 3B, Doublecortin-like and CAM kinase-like 2, doublecortin-like kinase 2CLICK2, EC 2.7.11, MGC45428, serine/threonine-protein kinase DCLK2 | |
Rabbit | |
85 kDa | |
0.1 mg | |
Signal Transduction | |
166614 | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
1 mg/mL | |
Western Blot 1-2 ug/ml, ELISA 1:100-1:2000, Immunohistochemistry 5 ug/ml, Immunohistochemistry-Paraffin 5 ug/ml | |
NP_001035350 | |
DCLK2 | |
Antibody was raised against a 14 amino acid synthetic peptide near the carboxy terminus of human DCLK2. The immunogen is located within the last 50 amino acids of DCLK2. Amino Acid Squence: PGGERAGTWRRHRD | |
Affinity Purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction