Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DCAMKL2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA582538
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA582538 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA582538 Supplier Invitrogen™ Supplier No. PA582538
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ENNMQAEVTG KLKQHFNNAL PKQNSTTTGV SVIMNTALDK EGQIFCSKHC QDSGRPGMEP ISPVPPSVEE IPVPGEAVPA PTPPESPTPH CPPAAPGGER AG Highest antigen sequence indentity to the following orthologs: Rat - 67%, Mouse - 74%.

DCAMKL2 is a serine/threonine kinase involved in signal transduction and cellular communication, and is the human homolog of a rat protein kinase expressed in cerebrocortical lamination that binds microtubules. The microtubule-polymerizing activity of the encoded protein is independent of its protein kinase activity. Mouse studies show that the DCX gene, another family member, and this gene share function in the establishment of hippocampal organization and that their absence results in a severe epileptic phenotype and lethality, as described in human patients with lissencephaly. Multiple alternatively spliced transcript variants have been identified.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DCAMKL2
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene DCLK2
Gene Accession No. Q8N568
Gene Alias 6330415M09Rik; AU044875; CaMK-like CREB regulatory kinase 2; CL2; CLICK2; Click-II; CLIK2; DCAMKL2; DCDC3; DCDC3B; DCK2; DCLK2; doublecortin and CaM kinase-like 2; doublecortin domain-containing protein 3B; doublecortin kinase 2; doublecortin like kinase 2; doublecortin-like and CAM kinase-like 2; Doublecortin-like kinase 2; RGD1308384; Serine/threonine-protein kinase DCLK2
Gene Symbols DCLK2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human DCLK2. Recombinant protein control fragment (Product #RP-91456).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 166614
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.