Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DCAMKL2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA520910
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA520910 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA520910 Supplier Invitrogen™ Supplier No. PA520910
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

A suggested positive control is rat brain tissue lysate. PA5-20910 can be used with blocking peptide PEP-1024.

DCAMKL2 is a serine/threonine kinase involved in signal transduction and cellular communication, and is the human homolog of a rat protein kinase expressed in cerebrocortical lamination that binds microtubules. The microtubule-polymerizing activity of the encoded protein is independent of its protein kinase activity. Mouse studies show that the DCX gene, another family member, and this gene share function in the establishment of hippocampal organization and that their absence results in a severe epileptic phenotype and lethality, as described in human patients with lissencephaly. Multiple alternatively spliced transcript variants have been identified.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DCAMKL2
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene DCLK2
Gene Accession No. Q5MPA9, Q6PGN3, Q8N568
Gene Alias 6330415M09Rik; AU044875; CaMK-like CREB regulatory kinase 2; CL2; CLICK2; Click-II; CLIK2; DCAMKL2; DCDC3; DCDC3B; DCK2; DCLK2; doublecortin and CaM kinase-like 2; doublecortin domain-containing protein 3B; doublecortin kinase 2; doublecortin like kinase 2; doublecortin-like and CAM kinase-like 2; Doublecortin-like kinase 2; RGD1308384; Serine/threonine-protein kinase DCLK2
Gene Symbols DCLK2
Host Species Rabbit
Immunogen A 14 amino acid peptide near the carboxy terminus of human DCLK2.
Purification Method Antigen Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 166614, 310698, 70762
Target Species Human, Mouse, Rat
Content And Storage Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.