Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DCANP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA572808
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA572808 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA572808 Supplier Invitrogen™ Supplier No. PA572808
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

DCANP1 Polyclonal Antibody for Western Blot, IHC (P), ELISA

Binds with and transactivates the corticotropin-releasing hormone (CRH) promoter.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DCANP1
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene DCANP1
Gene Accession No. Q8TF63
Gene Alias C5orf20; DCANP1; DCNP1; dendritic cell associated nuclear protein; dendritic cell nuclear protein 1; Dendritic cell nuclear protein-1; dendritic cell-associated nuclear protein
Gene Symbols DCANP1
Host Species Rabbit
Immunogen DCNP1 antibody for an 18 amino acid peptide near the amino terminus of human DCNP1. The immunogen is within the first 50 amino acids of DCNP1.
Purification Method Antigen Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 140947
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.