Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DCBLD2/ESDN Antibody, Allophycocyanin, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP300160APC
Description
DCBLD2/ESDN Polyclonal antibody specifically detects DCBLD2/ESDN in Human samples. It is validated for ELISASpecifications
DCBLD2/ESDN | |
Polyclonal | |
ELISA | |
CLCP11700055P21Rik, CUB, LCCL and coagulation factor V/VIII-homology domains protein 1, discoidin, CUB and LCCL domain containing 2, discoidin, CUB and LCCL domain-containing protein 2, Endothelial and smooth muscle cell-derived neuropilin-like protein, ESDNcoagulation factor V/VIII-homology domains protein 1 | |
Produced in rabbits immunized with purified, recombinant Human DCBLD2/ESDN (Accession#: NP_563615.3; Met1-Ala482) | |
0.1 mL | |
Signal Transduction | |
131566 | |
Store at 4C in the dark. | |
IgG |
ELISA | |
APC | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction