Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DCBLD2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5110820
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5110820 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5110820 Supplier Invitrogen™ Supplier No. PA5110820
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: AWHWRNRKKK TEGTYDLPYW DRAGWWKGMK QFLPAKAVDH EETPVRYSSS EVNHLSPREV TTVLQADSAE YAQPLVGGIV GTLHQRSTFK PEEGKEAGYA DLDPYNSPGQ EVYHAYAEPL PITGPEYATP IIMDMSGHPT TSVGQPST Highest antigen sequence indentity to the following orthologs: Mouse - 92%, Rat - 89%.

This protein is a key enzyme in the catabolic pathway of odd-chain fatty acids, isoleucine, threonine, methionine, and valine. It is probably a dodecamer composed of six biotin-containing alpha subunits and six beta subunits. The protein is located in the mitochondrial matrix. Defects in PCCA are the cause o fpropionic acidemia type I (PA-1) [MIM:606054]. PA-1 is a life-threatening disease characterized by episodic vomiting, lethargy and ketosis, neutropenia, periodic thrombocytopenia, hypogammaglobulinemia, develop-mental retardation, and intolerance to protein.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DCBLD2
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.10 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene DCBLD2
Gene Accession No. Q96PD2
Gene Alias 1700055P21Rik; AW146002; CLCP1; coagulation factor V/VIII-homology domains protein 1; CUB, LCCL and coagulation factor V/VIII-homology domains protein 1; DCBLD2; discoidin; discoidin, CUB and LCCL domain containing 2; discoidin, CUB and LCCL domain-containing protein 2; Endothelial and smooth muscle cell-derived neuropilin-like protein; ESDN
Gene Symbols DCBLD2
Host Species Rabbit
Immunogen Recombinant Protein corresonding to Human DCBLD2. Recombinant protein control fragment (Product #RP-89964).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 131566
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.