Learn More
DCBLD2 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A304454AT

Description
The recommended shelf life for this product is 1 year from date of receipt.
DCBLD2, otherwise known as ESDN (endothelial and smooth muscle cell-derived neuropilin-like molecule) is a novel type-I transmembrane protein with the longest cleavable secretory signal sequence among eukaryotes. It is expressed in various tissues; particularly highly expressed in cultured vascular smooth muscle cells. DCBLD2 is considered to play a role in regulation of vascular cell growth and may have a wide variety of functions in other tissues including the nervous system, like neuropilins.Specifications
DCBLD2 | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Q91ZV3, Q96PD2 | |
Dcbld2 | |
Between 725 and 775 | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
DCBLD2 | |
CLCP1, ESDN | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
131566, 73379 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.