Learn More
Invitrogen™ DCC Monoclonal Antibody (531505)
Description
Reconstitute at 0.2 mg/mL in sterile PBS.
Specifications
Specifications
| Antigen | DCC |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | 531505 |
| Conjugate | Unconjugated |
| Formulation | PBS with No Preservative |
| Gene | DCC |
| Gene Accession No. | P43146 |
| Gene Alias | C030036D22Rik; Colorectal cancer suppressor; colorectal tumor suppressor; CRC18; CRCR1; Dcc; DCC netrin 1 receptor; deleted in colorectal cancer protein; deleted in colorectal carcinoma; IGDCC1; Immunoglobulin superfamily DCC subclass member 1; immunoglobulin superfamily, DCC subclass, member 1; MRMV1; Netrin receptor DCC; NTN1R1; putative colorectal tumor suppressor; tumor suppressor protein DCC |
| Gene Symbols | DCC |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.