Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DCC Polyclonal Antibody, Abbomax

Catalog No. 502203980
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-3980 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-3980 Supplier Abbomax Inc Supplier No. 601460
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive control: Brain tissue Cellular location: Cell Membrane.

DCC Polyclonal antibody specifically detects DCC in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen DCC
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with proprietary stabilizer and 0.01% sodium azide
Gene DCC
Gene Accession No. P43146
Gene Alias C030036D22Rik; Colorectal cancer suppressor; colorectal tumor suppressor; CRC18; CRCR1; Dcc; DCC netrin 1 receptor; deleted in colorectal cancer protein; deleted in colorectal carcinoma; IGDCC1; Immunoglobulin superfamily DCC subclass member 1; immunoglobulin superfamily, DCC subclass, member 1; MRMV1; Netrin receptor DCC; NTN1R1; putative colorectal tumor suppressor; tumor suppressor protein DCC
Host Species Rabbit
Immunogen A synthetic peptide corresponding to the C-terminus (within 1380-1447aa) of Human DCC protein
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1630
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.