Learn More
Description
This antibody raised against a synthesized polypeptide of human DCC protein recognizes endogenous DCC with an apparent molecular weight of 180 kDa. As the predicted molecular mass of DCC is 158 kDa, the slower mobility is likely attributable to post-trans
Specifications
Specifications
| Antigen | DCC |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.43 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 |
| Gene | DCC |
| Gene Accession No. | P43146, P70211, Q63155 |
| Gene Alias | Colorectal cancer suppressor, CRC18, CRCR1, DCC, IGDCC1, Netrin receptor DCC, Tumor suppressor protein DCC |
| Gene Symbols | DCC |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
