Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DCD Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596623
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596623 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596623 Supplier Invitrogen™ Supplier No. PA596623
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: YDPEAASAPG SGNPCHEASA AQKENAGEDP GLARQAPKPR KQRSSLLEKG LDGAKKAVGG LGKLGKDAVE DLESVGKGAV HDVKDVLDSV L; Positive Samples: Mouse skin.

DCD is a secreted protein that is subsequently processed into mature peptides of distinct biological activities. The C-terminal peptide is constitutively expressed in sweat and has antibacterial and antifungal activities. The N-terminal peptide, also known as diffusible survival evasion peptide, promotes neural cell survival under conditions of severe oxidative stress. A glycosylated form of the N-terminal peptide may be associated with cachexia (muscle wasting) in cancer patients.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DCD
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.7 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene DCD
Gene Accession No. P81605
Gene Alias AIDD; DCD; DCD-1; Dermcidin; diffusible survival/evasion peptide; DSEP; HCAP; PIF; Preproteolysin; proteolysis inducing factor; survival promoting peptide; Survival-promoting peptide
Gene Symbols DCD
Host Species Rabbit
Immunogen Recombinant protein (or fragment).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 117159
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.