Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DCDC2B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA561101
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA561101 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA561101 Supplier Invitrogen™ Supplier No. PA561101
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VVYRNGDPFF PGSQLVVTQR RFPTMEAFLC EVTSAVQAPL AVRALYTPCH GHPVTNLADL KNRGQYVAAG FERFHK Highest antigen sequence identity to the following orthologs: Mouse - 84%, Rat - 88%.

This gene encodes a member of the doublecortin family. The protein encoded by this gene contains two doublecortin domains. The doublecortin domain has been demonstrated to bind tubulin and enhance microtubule polymerization.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DCDC2B
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene DCDC2B
Gene Accession No. A2VCK2
Gene Alias DCDC2B; doublecortin domain containing 2B; doublecortin domain-containing protein 2B
Gene Symbols DCDC2B
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human DCDC2B. Recombinant protein control fragment (Product #RP-94577).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 149069
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.