Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ DCEBIO
GREENER_CHOICESDP

Catalog No. 142250 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-225-0 50 mg
14-221-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 14-225-0 Supplier Tocris Bioscience™ Supplier No. 1422/50
Add to Cart
Add to Cart
This item is not returnable. View return policy

Activates Cl- conductance and hKCa3.1 channels

DCEBIO is a stimulates Cl- secretion via the activation of hKCa3.1 (IK1) potassium channels and an apical Cl- conductance carried by the CFTR channel. More potent than its analog 1-EBIO (Cat. No. 1041).

Specifications

Chemical Name or Material 5,6-Dichloro-1-ethyl-1,3-dihydro-2H-benzimidazol-2-one
CAS 60563-36-2
Assay Percent Range >99%
Quantity 50 mg
Solubility Soluble to 100mM in DMSO
Molecular Formula C9H8Cl2N2O
Formula Weight Observed MW: 231.08

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.