Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ DCG IV
GREENER_CHOICESDP

Catalog No. 097550 Shop All Novus Products
Change view
Click to view available options
Quantity:
1 mg
10 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
09-755-0 50 mg
09-751-0 10 mg
09-751-R 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 09-755-0 Supplier R&D Systems™ Supplier No. 0975/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Highly potent group II mGlu agonist; also NMDA agonist

DCG IV is a presynaptic depressant; highly potent agonist for group II mGlu receptors.

Specifications

Chemical Name or Material (2 S,2 'R,3 'R)-2-(2 ',3 '-Dicarboxycyclopropyl)glycine
CAS 147782-19-2
Assay Percent Range >98%
Quantity 50 mg
Solubility Soluble to 100mM in water
Product Line Tocris
Target Group II mGluR Agonists
Molecular Formula C7H9NO6
Formula Weight Observed MW: 203.15
Purity 98%

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.