Learn More
Invitrogen™ DCI Polyclonal Antibody

Description
Positive test controls include: BxPC-3, HeLa, 22Rv1, Mouse heart, Mouse liver. The target is usually found in the following locations: Mitochondrion matrix. Immunogen sequence: KNPPVNSLSL EFLTELVISL EKLENDKSFR GVILTSDRPG VFSAGLDLTE MCGRSPAHYA GYWKAVQELW LRLYQSNLVL VSAINGACPA GGCLVALTCD YRILADNPRY CIGLNETQLG IIAPFWLKDT LENTIGHRAA ERALQLGLLF PPAEALQVGI VDQVVPEEQV QSTALSAIAQ WMAIPDHARQ LTKAMMRKAT ASRLVTQRDA DVQNFVSFIS KDSIQKSLQM YLERLKEEKG.
Specifications
Specifications
| Antigen | DCI |
| Applications | ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 2.73 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | Eci1 |
| Gene Accession No. | P42125, P42126 |
| Gene Alias | 3,2 trans-enoyl-coenyme A isomerase; 3,2 trans-enoyl-Coenzyme A isomerase; 3,2-trans-enoyl-CoA isomerase; 3,2-trans-enoyl-CoA isomerase, mitochondrial; ACBD2; acetylene-allene isomerase; D3,D2-enoyl-CoA isomerase; Dci; Delta(3),Delta(2)-enoyl-CoA isomerase; dodecenoyl-CoA de; dodecenoyl-CoA delta isomerase (3,2 trans-enoyl-CoA isomerase); dodecenoyl-CoA isomerase; dodecenoyl-Coenzyme A delta isomerase (3,2 trans-enoyl-Coenyme A isomerase); dodecenoyl-Coenzyme A delta isomerase (3,2 trans-enoyl-Coenzyme A isomerase); DRS 1; DRS1; eci; ECI1; enoyl-CoA delta isomerase 1; enoyl-CoA delta isomerase 1, mitochondrial; enoyl-Coenzyme A delta isomerase 1; HCA88; Peroxisomal 3 |
| Gene Symbols | Eci1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.