Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DCIR/CLEC4A Antibody (MM0237-5P20) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP212257
Description
DCIR/CLEC4A Monoclonal specifically detects DCIR/CLEC4A in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.Specifications
DCIR/CLEC4A | |
Monoclonal | |
LYOPH | |
Western Blot 1:500-1:1000, Flow Cytometry 1:10-1:1000, CyTOF-ready | |
Q9UMR7 | |
CLEC4A | |
Human recombinant DCIR/CLEC4A (extracellular domain) | |
0.1 mg | |
Immunology, Innate Immunity | |
50856 | |
Human | |
Purified |
Western Blot, Flow Cytometry, CyTOF | |
MM0237-5P20 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS. 0.025 mg size is provided in liquid form, PBS with No Preservative | |
CLECSF6C-type lectin superfamily member 6, C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamilymember 6, C-type lectin domain family 4 member A, C-type lectin domain family 4, member A, DCIRLLIR, DDB27C-type lectin DDB27, Dendritic cell immunoreceptor, Lectin-like immunoreceptor | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Reconstitute with sterilized PBS to a final concentration of 0.5 mg/ml. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction