Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DCK Antibody (10412), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP243639
Description
Ensure accurate, reproducible results in Western Blot, Immunohistochemistry (Paraffin), Immunoprecipitation
DCK Monoclonal specifically detects DCK in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.Specifications
DCK | |
Monoclonal | |
Unconjugated | |
PBS with No Preservative | |
DCK | |
Recombinant protein encompassing a sequence within the center region of human DCK. The exact sequence is proprietary. | |
Protein A purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
10412 | |
Western Blot 1:1000-1:10000, Immunohistochemistry 1:100-1:1000, Immunocytochemistry/Immunofluorescence 1:100-1:1000, Immunoprecipitation 1:100-1:500, Immunohistochemistry-Paraffin 1:100-1:1000 | |
dCK, deoxycytidine kinase, EC 2.7.1, EC 2.7.1.74, MGC117410, MGC138632 | |
Mouse | |
31 kDa | |
0.1 mL | |
Proteases & Other Enzymes | |
1633 | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction