Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DCK Antibody (OTI3F5), DyLight 680, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP270555FR
Description
DCK Monoclonal specifically detects DCK in Human, Mouse, Rat, Monkey samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
DCK | |
Monoclonal | |
DyLight 680 | |
PBS with 0.05% Sodium Azide | |
DCK | |
Full length recombinant protein of human DCK (NP_000779) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat, Monkey | |
IgG2a |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
OTI3F5 | |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
dCK, deoxycytidine kinase, EC 2.7.1, EC 2.7.1.74, MGC117410, MGC138632 | |
Mouse | |
Affinity Purified | |
Proteases & Other Enzymes | |
1633.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction