Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ DCK Monoclonal Antibody (2F10)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA518705
Description
Peptide Sequence: WMNNQFGQSL ELDGIIYLQA TPETCLHRIY LRGRNEEQGI PLEYLEKLHY KHESWLLHRT LKTNFDYLQE VPILTLDVNE DFKDKYESLV EKVKEFLSTL.
Deoxycytidine kinase (DCK) is required for the phosphorylation of several deoxyribonucleosides and their nucleoside analogs. Deficiency of DCK is associated with resistance to antiviral and anticancer chemotherapeutic agents. Conversely, increased deoxycytidine kinase activity is associated with increased activation of these compounds to cytotoxic nucleoside triphosphate derivatives. DCK is clinically important because of its relationship to drug resistance and sensitivity.
Specifications
DCK | |
Monoclonal | |
0.5 mg/mL | |
PBS with no preservative; pH 7.4 | |
P27707 | |
DCK | |
DCK (NP_000779, 161 a.a. ~ 260 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2b κ |
ELISA, Western Blot, Immunocytochemistry | |
2F10 | |
Unconjugated | |
DCK | |
DCK; Deoxycytidine kinase; deoxynucleoside kinase | |
Mouse | |
Affinity chromatography | |
RUO | |
1633 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction