Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DCK Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA584564
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA584564 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA584564 Supplier Invitrogen™ Supplier No. PA584564
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VNILKQLCED WEVVPEPVAR WCNVQSTQDE FEELTMSQKN GGNVLQMMYE KPERWSFTFQ TYACLSRIRA QLASLNGKLK D Highest antigen sequence indentity to the following orthologs: Mouse - 93%, Rat - 91%.

Deoxycytidine kinase (DCK) is required for the phosphorylation of several deoxyribonucleosides and their nucleoside analogs. Deficiency of DCK is associated with resistance to antiviral and anticancer chemotherapeutic agents. Conversely, increased deoxycytidine kinase activity is associated with increased activation of these compounds to cytotoxic nucleoside triphosphate derivatives. DCK is clinically important because of its relationship to drug resistance and sensitivity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DCK
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene DCK
Gene Accession No. P27707
Gene Alias DCK; Deoxycytidine kinase; deoxynucleoside kinase
Gene Symbols DCK
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human DCK. Recombinant protein control fragment (Product #RP-103469).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1633
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.