Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DCLK1 Polyclonal Antibody
DCLK1 Polyclonal Antibody
Supplier: Thermo Scientific PA553361
Description
DCLK1 Polyclonal Antibody for IHC

Specifications
DCLK1 | |
Polyclonal | |
Unconjugated | |
DCLK1 | |
1700113D08Rik; 2810480F11Rik; AI836758; Ania4; Calcium/calmodulin-dependent protein kinase type I-like CPG16; CL1; CLICK1; Click-I; CLICK-I beta; Cpg16; Dcamkl1; DCDC3A; Dcl; Dclk; Dclk1; double cortin and calcium/calmodulin-dependent protein kinase-like 1; doublecortin and calcium/calmodulin-dependent protein kinase-like 1; doublecortin domain-containing protein 3A; doublecortin like kinase 1; doublecortin-like and CAM kinase-like 1; Doublecortin-like kinase 1; KIAA0369; mKIAA0369; RP11-113P14.1; Serine/threonine-protein kinase DCLK1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
9201 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O15075 | |
DCLK1 | |
Recombinant protein corresponding to Human DCAMKL1. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction