Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DCLK1 Polyclonal Antibody, Invitrogen™

Catalog No. PIPA595770
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA595770 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA595770 Supplier Thermo Scientific Supplier No. PA595770

Rabbit Polyclonal Antibody

Immunogen sequence: VDVDQRFSAV QVLEHPWVND DGLPENEHQL SVAGKIKKHF NTGPKPNSTA AGVSVIALDH GFTIKRSGSL DYYQQPGMYW IRPPLLIRRG RFSDEDATRM; Positive Samples: HepG2, A-549, 22Rv1, Mouse heart

DCAMKL1 (DCLK1) serine/threonine kinase or DCLK1 or doublecortin-like kinase 1 contains two N-terminal doublecortin domains (which bind microtubules and regulate microtubule polymerization), a C-terminal serine/threonine protein kinase domain (which shows substantial homology to Ca2+/calmodulin-dependent protein kinase), and a serine/proline-rich domain in between the doublecortin and the protein kinase domains (which mediates multiple protein-protein interactions). DCAMKL1 is a microtubule-associated kinase that can undergo autophosphorylation. DCAMKL1 has microtubule-polymerizing activity that is independent of its protein kinase activity. DCAMKL1 is involved in several different cellular processes, including neuronal migration, retrograde transport, neuronal apoptosis and neurogenesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DCLK1
Applications Western Blot
Classification Polyclonal
Concentration 2.29 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal, pH 7.3
Gene DCLK1
Gene Accession No. O15075, Q9JLM8
Gene Alias 1700113D08Rik, 2810480F11Rik, AI836758, Ania4, Calcium/calmodulin-dependent protein kinase type I-like CPG16, CL1, CLICK1, Click-I, CLICK-I beta, Cpg16, Dcamkl1, DCDC3A, Dcl, Dclk, Dclk1, double cortin and calcium/calmodulin-dependent protein kinase-like 1, doublecortin and calcium/calmodulin-dependent protein kinase-like 1, doublecortin domain-containing protein 3A, doublecortin like kinase 1, doublecortin-like and CAM kinase-like 1, Doublecortin-like kinase 1, KIAA0369, mKIAA0369, RP11-113P14.1, Serine/threonine-protein kinase DCLK1
Gene Symbols DCLK1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 630-729 of human DCAMKL1/DCAMKL1/DCLK1 (NP_0047251)
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13175, 9201
Target Species Human, Mouse
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.