Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DCLK1 Rabbit anti-Human, Mouse, Rat, Clone: 8C8T3, Novus Biologicals™

Rabbit Monoclonal Antibody
$205.50 - $492.50
Specifications
Antigen | DCLK1 |
---|---|
Clone | 8C8T3 |
Applications | Western Blot |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
DCLK1 Monoclonal antibody specifically detects DCLK1 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
DCLK1 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Protein Kinase | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
9201 | |
IgG | |
Affinity purified |
8C8T3 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
DCAMKL1CLICK1, DCDC3ACL1, DCLK, doublecortin and CaM kinase-like 1, Doublecortin domain-containing protein 3A, doublecortin-like kinase 1Doublecortin-like and CAM kinase-like 1, EC 2.7.11, KIAA0369EC 2.7.11.1, serine/threonine-protein kinase DCLK1 | |
A synthetic peptide corresponding to a sequence within amino acids 600-700 of human DCLK1 (O15075). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title