Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DCLK1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
| Antigen | DCLK1 |
|---|---|
| Concentration | 0.23 mg/mL |
| Applications | Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-3553
![]()
|
Proteintech Group Inc
216991AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
Eukaryotic translation initiation factor 4 gamma 2 (elF4G or DAP5) plays a role in the switch from cap-dependent to IRES-mediated translation during mitosis, apoptosis, and viral infections. eIF4G2 is a ubiquitously expressed member of the translation initiation factor eIF4G family that lacks the EIF4E binding site.Specifications
| DCLK1 | |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| O08875, O15075, Q9JLM8 | |
| 13175, 83825, 9201 | |
| DCLK1 Fusion Protein Ag16292 | |
| Primary | |
| -20°C | |
| DCLK1 |
| 0.23 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| DCAMKL1, DCDC3A, DCLK, DCLK1, doublecortin like kinase 1, KIAA0369 | |
| DCLK1 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title