Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DCLK1-S Rabbit anti-Human, Unconjugated, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma ABC138925UL
Description
Anti-DCLK1-S, Cat. No. ABC1389, is a rabbit polyclonal antibody that specifically detects the short version of Serine/threonine-protein kinase DCLK1. It has been tested for use in Immunohistochemistry and Western Blotting.
Specifications
DCLK1-S | |
Polyclonal | |
Unconjugated | |
Serine/threonine-protein kinase DCLK1-S;EC: 2.7.11.1;Doublecortin domain-containing protein 3A-short;Doublecortin-like and CAM kinase-like 1;Doublecortin-like kinase 1;PS41014 | |
Rabbit | |
∽55 kDa observed. 47.60 kDa calculated. Uncharacterized bands may be observed in some lysate(s). | |
25 μL | |
Apoptosis & Cancer | |
This rabbit polyclonal antibody detects the short version of Serine/threonine-protein kinase DCLK1 (DCLK1-S). It does not react with long isoforms. It targets an epitope within six amino acids from the N-terminus region. | |
Stable for 1 year at 2-8°C from date of receipt. | |
Purified |
Immunohistochemistry, Western Blot | |
Please refer to lot specific datasheet. | |
Purified rabbit polyclonal antibody in PBS with 0.05% sodium azide. | |
DCLK1;DCAMKL1;;DCDC3A;KIAA0369;DCLK1 | |
A multimeric scaffold of the 6 amino acids from the N-terminal region of the short version of human DCLK1 (DCLK1-S). | |
Affinity Purified | |
RUO | |
Primary | |
Human | |
Antibodies | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction