Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DCLK3 Antibody, mFluor Violet 450 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP177129MFV450
Description
DCLK3 Polyclonal antibody specifically detects DCLK3 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
DCLK3 | |
Polyclonal | |
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin | |
CLICK-I, DCAMKL3, DCDC3CCLR, doublecortin and CaM kinase-like 3, Doublecortin domain-containing protein 3C, Doublecortin-like and CAM kinase-like 3, doublecortin-like kinase 3EC 2.7.11.1, EC 2.7.11, II-related, KIAA1765DCK3, serine/threonine-protein kinase DCLK3 | |
Antibody was raised against a 14 amino acid synthetic peptide near the amino terminus of human DCLK3. The immunogen is located within the last 50 amino acids of DCLK3. Amino Acid Squence: SEGHFRSQHKRVVE | |
0.1 mL | |
Neuroscience | |
85443 | |
Store at 4C in the dark. | |
IgG |
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin) | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Rabbit | |
Peptide affinity purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction