Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DCLK3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA5144847

Description
Immunogen sequence: PRNPTQELRR PSKSMDKKED RGPEDQESHA QGAAKAKKDL VEVLPVTEEG LREVKKDTRP MSRSKHGGWL LREHQAGFEK LRRTRGEEKE AEKEKKPCMS GGRRMTLRDD QPAKL Antigen sequence identity: Mouse 53%; Rat 56%
DCLK3 Polyclonal antibody specifically detects DCLK3 in Human samples. It is validated for Immunocytochemistry
Specifications
DCLK3 | |
Polyclonal | |
Unconjugated | |
DCLK3 | |
BC056929, C730036H08, CLICK-I and II-related, Click-I, -II related, CLICK-I,II-related, CLR, Dcamkl3, DCDC3C, DCK3, Dclk3, doublecortin and CaM kinase-like 3, doublecortin domain-containing protein 3C, doublecortin like kinase 3, doublecortin-like and CAM kinase-like 3, Doublecortin-like kinase 3, hCG_16695, KIAA1765, RGD1309232, serine/threonine-protein kinase DCLK3 | |
Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence. Antigen range: 207-321. | |
100 μL | |
Primary | |
Human | |
Liquid |
Immunocytochemistry | |
0.6 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9C098 | |
Rabbit | |
Affinity chromatography | |
RUO | |
85443 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction