Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DCLK3 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA520911

Catalog No. PIPA520911


Only null left
Add to Cart

Description

Description

A suggested positive control is K562 cell lysate. PA5-20911 can be used with blocking peptide PEP-1025.

DCLK3 is one of three doublecortin-like kinases similar to the Ca2+/calmodulin-dependent protein kinase (CaMK) family. DCLK3 mRNA, like that of the homologous DCLK1 and DCLK3, is highly expressed in adult brain, but only DCLK3 transcripts are present in liver and kidney, suggesting that DCLK3 may play other roles than in cortical development. The DCLK proteins are homologous to Doublecortin (DCX), a protein that is mutated in X-linked human lissencephaly. In mouse models where the DCX gene has been disrupted, DCLK1 expression increases slightly and appears to compensate for the loss of DCX, as mice mutant for both DCX and DCLK1 show a severe phenotype including perinatal lethality, disorganized neocortical layering, and profound hippocampal cytoarchitectural disorganization.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

DCLK3
Polyclonal
Unconjugated
DCLK3
BC056929; C730036H08; CLICK-I and II-related; Click-I, -II related; CLICK-I,II-related; CLR; Dcamkl3; DCDC3C; DCK3; Dclk3; doublecortin and CaM kinase-like 3; doublecortin domain-containing protein 3C; doublecortin like kinase 3; doublecortin-like and CAM kinase-like 3; Doublecortin-like kinase 3; hCG_16695; KIAA1765; RGD1309232; serine/threonine-protein kinase DCLK3
Rabbit
Antigen affinity chromatography
RUO
245038, 85443
Maintain refrigerated at 2°C to 8°C for up to 3 months. For long term storage store at -20°C
Liquid
Immunohistochemistry, Western Blot
1 mg/mL
PBS with 0.02% sodium azide
Q8BWQ5, Q9C098
DCLK3
A 14 amino acid peptide near the amino terminus of human DCLK3.
100 μg
Primary
Human, Mouse
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

WARNING: Cancer - www.P65Warnings.ca.gov
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.