Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DCLRE1A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA557868
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA557868 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA557868 Supplier Invitrogen™ Supplier No. PA557868
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TTPGKLCRTQ KSQHVSPKIR PVYDGYCPNC QMPFSSLIGQ TPRWHVFECL DSPPRSETEC PDGLLCTSTI PFHYKRYTHF LLAQSRAG Highest antigen sequence identity to the following orthologs: Mouse - 76%, Rat - 74%.

May be required for DNA interstrand cross-link repair. Also required for checkpoint mediated cell cycle arrest in early prophase in response to mitotic spindle poisons.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DCLRE1A
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene DCLRE1A
Gene Accession No. Q6PJP8
Gene Alias DCLRE1A; DNA cross-link repair 1A; DNA cross-link repair 1A protein; DNA-crosslink repair gene SNM1; hSNM1; hSNM1A; KIAA0086; PSO2; SNM1; SNM1 homolog A; SNM1A
Gene Symbols DCLRE1A
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human DCLRE1A. Recombinant protein control fragment (Product #RP-96587).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9937
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.