Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ DCLRE1A Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA571370
Description
DCLRE1A Polyclonal Antibody for Western Blot
May be required for DNA interstrand cross-link repair. Also required for checkpoint mediated cell cycle arrest in early prophase in response to mitotic spindle poisons.
Specifications
DCLRE1A | |
Polyclonal | |
Unconjugated | |
DCLRE1A | |
DCLRE1A; DNA cross-link repair 1A; DNA cross-link repair 1A protein; DNA-crosslink repair gene SNM1; hSNM1; hSNM1A; KIAA0086; PSO2; SNM1; SNM1 homolog A; SNM1A | |
Rabbit | |
Affinity chromatography | |
RUO | |
9937 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q6PJP8 | |
DCLRE1A | |
synthetic peptide directed towards the N terminal region of human DCLRE1A. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction