Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DCLRE1B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA593234
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA593234 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA593234 Supplier Invitrogen™ Supplier No. PA593234
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VQQYMSSSSR KPSLLWLLER RLKRPRTQGV VFESPEESAD QSQADRDSKK AKKEKLSPWP ADLEKQPSHH PLRIKKQLFP DLYSKEWNKA VPFCESQKRV TMLTAPLGFS VHLRSTDEEF ISQKTREEIG LGSPLVPMGD DDGGPEATGN QSAWMGHGSP LSHSSKGTPL LATEFRGLAL KYLLTPVNFF QAGYSSRRFD QQVEKYHKPC; Positive Samples: THP-1; Cellular Location: Chromosome, Cytoplasm, Nucleus, centrosome, cytoskeleton, microtubule organizing center, telomere.

DNA interstrand cross-links prevent strand separation, thereby physically blocking transcription, replication, and segregation of DNA. DCLRE1B is one of several evolutionarily conserved genes involved in repair of interstrand cross-links.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DCLRE1B
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.63 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Dclre1b
Gene Accession No. Q8C7W7, Q9H816
Gene Alias 5' exonuclease Apollo; AI452214; APOLLO; DCLRE1B; DNA cross-link repair 1B; DNA cross-link repair 1B (PSO2 homolog, S. cerevisiae); DNA cross-link repair 1B protein; DNA cross-link repair 1B, PSO2 homolog (S. cerevisiae); mSNM1B; PSO2 homolog; SNM1 homolog B; Snm1b; SNMIB
Gene Symbols Dclre1b
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 323-532 of human DCLRE1B (NP_0737471).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 140917, 64858
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.