Learn More
Invitrogen™ DCLRE1B Polyclonal Antibody

Description
Antibody detects endogenous levels of total DCLRE1B.
Specifications
Specifications
| Antigen | DCLRE1B |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | Dclre1b |
| Gene Accession No. | Q9H816 |
| Gene Alias | 5' exonuclease Apollo; AI452214; APOLLO; DCLRE1B; DNA cross-link repair 1B; DNA cross-link repair 1B (PSO2 homolog, S. cerevisiae); DNA cross-link repair 1B protein; DNA cross-link repair 1B, PSO2 homolog (S. cerevisiae); mSNM1B; PSO2 homolog; SNM1 homolog B; Snm1b; SNMIB |
| Gene Symbols | Dclre1b |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.