Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DCLREC1C/Artemis Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501982317
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-2317 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-2317 Supplier Bioss Supplier No. BS12533R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The protein encoded by this gene is a receptor for aryl hydrocarbons and a ligand-activated transcription factor. The encoded protein is found in the cytoplasm as part of a multiprotein complex, but upon binding of ligand is transported to the nucleus. This protein can regulate the expression of many xenobiotic metabolizing enzymes. Also, the encoded protein can bind specifically to and inhibit the activity of hepatitis B virus.

Specifications

Antigen DCLREC1C/Artemis
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene DCLRE1C
Gene Accession No. Q96SD1
Gene Alias 9930121L06Rik; AI661365; Art; ARTEMIS; ASCID; A-SCID; Dclre1c; DCLRE1C protein; DCLREC1C; DNA cross-link repair 1A, PSO2 homolog; DNA cross-link repair 1C; DNA cross-link repair 1C (PSO2 homolog, S. cerevisiae); DNA cross-link repair 1C protein; DNA cross-link repair 1C, PSO2 homolog (S. cerevisiae); FLJ11360; hSNM1C; mArt; Protein artemis; Protein A-SCID; RS SCID; RS-SCID; SCIDA; severe combined immunodeficiency, type a (Athabascan); SNM1 homolog C; SNM1C; SNM1C antibody; Snm1l; SNM1-like protein; tein
Gene Symbols DCLRE1C
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human DCLREC1C/Artemis.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64421
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.