Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ DCM Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5144467
Description
DCM Polyclonal Antibody for Western Blot, ELISA
This methylase recognizes the double-stranded sequence 5'-CCWGG-3', methylates C-2 on both strands.
Specifications
DCM | |
Polyclonal | |
Unconjugated | |
dcm | |
mec | |
Rabbit | |
Protein A/G | |
RUO | |
75205799 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Western Blot | |
2.18 mg/mL | |
PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4 | |
P0AED9 | |
dcm | |
Recombinant Escherichia coli (strain K12) DCM protein. | |
100 μL | |
Primary | |
Bacteria | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction