Learn More
Invitrogen™ DCP1A Polyclonal Antibody

Description
Positive test controls include: HepG2, Jurkat, Mouse brain, Rat brain. The target is usually found in the following locations: Cytoplasm, Nucleus, P-body. Immunogen sequence: NEKHAPTYTI PLSPVLSPTL PAEAPTAQVP PSLPRNSTMM QAVKTTPRQR SPLLNQPVPE LSHASLIANQ SPFRAPLNVT NTAGTSLPSV DLLQKLRLTP QHDQIQTQPL GKGAMVASFS PAAGQLATPE SFIEPPSKTA AARVAASASL SNMVLAPLQS MQQNQDPEVF VQPKVLSSAI PVAGAPLVTA TTTAVSSVLL APSVFQQTVT RSSDLERKAS SPSPLTIGTP ESQRKPSIIL SKSQLQDTLI HLIKNDSSFL STLHEVYLQV LTKNKDNHNL.
Specifications
Specifications
| Antigen | DCP1A |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Concentration | 1.23 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.09% sodium azide; pH 7.3 |
| Gene | Dcp1a |
| Gene Accession No. | Q91YD3, Q9NPI6 |
| Gene Alias | 1110066A22Rik; 4930568L04Rik; AU019772; D14Ertd817e; DCP1 decapping enzyme homolog A; DCP1 decapping enzyme homolog A (S. cerevisiae); DCP1 decapping enzyme-like protein A; DCP1A; decapping enzyme; decapping enzyme hDcp1a; decapping mRNA 1A; FLJ21691; HSA275986; MAD homolog 4 interacting transcription coactivator 1; MAD homolog 4-interacting transcription coactivator 1; Mitc1; mRNA-decapping enzyme 1A; Nbla00360; putative protein product of Nbla00360; RGD1561298; Smad4-interacting transcriptional co-activator; SMAD4IP1; SMIF; test2; transcription factor SMIF |
| Gene Symbols | Dcp1a |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.