Learn More
DCP1A Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A303592A

Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Mouse
Decapping is a key step in general and regulated mRNA decay. The protein encoded by this gene is a decapping enzyme. This protein and another decapping enzyme form a decapping complex, which interacts with the nonsense-mediated decay factor hUpf1 and may be recruited to mRNAs containing premature termination codons. This protein also participates in the TGF-beta signaling pathway.Specifications
DCP1A | |
Polyclonal | |
Unconjugated | |
DCP1A | |
DCP1A; decapping enzyme ; HSA275986; Nbla00360; SMAD4IP1; SMIF; Smad4-interacting transcriptional co-activator; decapping enzyme hDcp1a; putative protein product of Nbla00360; transcription factor SMIF | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
55802 | |
4° C | |
Liquid |
Immunoprecipitation, Western Blot | |
1 mg/ml | |
phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
Q9NPI6 | |
DCP1A | |
Between 532 and 582 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.