Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DCP1B Antibody, DyLight 755, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP204009IR
Description
DCP1B Polyclonal antibody specifically detects DCP1B in Human samples. It is validated for Western BlotSpecifications
DCP1B | |
Polyclonal | |
Western Blot | |
DCP1, DCP1 decapping enzyme homolog B (S. cerevisiae), decapping enzyme hDcp1b, EC 3.-, FLJ31638, hDcp1b, mRNA-decapping enzyme 1B | |
The immunogen recognized by this antibody maps to a region between residue 250 and 300 of human DCP1 Decapping Enzyme Homolog B using the numbering given in entry NP_689853.3 (GeneID 196513). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot | |
DyLight 755 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
196513 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction