Learn More
DCP1B Mouse anti-Human, Clone: OTI1G8, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF809931

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
DCP1B is a core component of the mRNA decapping complex, a key factor in the regulation of mRNA decay.Specifications
DCP1B | |
Monoclonal | |
Unconjugated | |
DCP1B | |
DCP1 | |
Mouse | |
Affinity Chromatography | |
RUO | |
196513 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI1G8 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q8IZD4 | |
DCP1B | |
Human recombit protein fragment corresponding to amino acids 167-473 of human DCP1B produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.